missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse CRYAB Protein
A cDNA sequence encoding the CRYAB was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11518-5ug
Additional Details : Weight : 0.01000kg
Specifications
CRYAB Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MDIAIHHPWI RRPFFPFHSP SRLFDQFFGE HLLESDLFST ATSLSPFYLR PPSFLRAPSW IDTGLSEMRL EKDRFSVNLD VKHFSPEELK VKVLGDVIEV HGKHEERQDE HGFISREFHR KYRIPADVDP LTITSSLSSD GVLTVNGPRK QVSGPERTIP ITREEKPAVA AAPKK | |
Greater than 95.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Mouse | |
Untagged | |
The Mouse CRYAB protein solution contains 20mM Tris-HCl buffer pH 8 and 10% glycerol. |