missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Clathrin light chain A Protein
A DNA sequence encoding the Mus musculus (Mouse) Clathrin light chain A, was expressed in the hosts and tags indicated.
£439.00 - £1230.00
Specifications
Name | Clathrin light chain A Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | Clta |
Protein Length | Full Length |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15968159
|
enQuireBio™
QP5841-MA-20UG |
20 μg |
£439.00
20µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15958159
|
enQuireBio™
QP5841-MA-100UG |
100 μg |
£1230.00
100µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15978159
|
enQuireBio™
QP5841-MA-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
Clathrin light chain A Protein | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
Research Use Only | |
Clta | |
Recombinant Protein | |
Mammal | |
MAELDPFGAP AGAPGGPALG NGVAGAGEED PAAAFLAQQE SEIAGIENDE AFAILDGGAP GRATRRAAGG PDAVDGVMNG EYYQESNGPT DSYAAISEVD RLQSEPESIR KWREEQTERL EALDANSRKQ EAEWKEKAIK ELEEWYARQD EQLQKTKANN RVADEAFYKQ PFADLIGYVA AEEAFVNDID ESSPGTEWER VARLCDFNPK SSKQAKDVSR MRSVLISLKQ APLVH | |
Greater than 90% as determined by SDS-PAGE. |