missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Chitinase domain-containing protein 1
A DNA sequence encoding the Mus musculus (Mouse) Chitinase domain-containing protein 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP10054-ba-20ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001136153.1 | |
Chitinase domain-containing protein 1 | |
Research Use Only | |
Chid1 | |
Recombinant Protein | |
Baculovirus | |
TLSKSDAKKA ASKMLLEKTQ FSDKPVQDRG LVVTDIKAED VVLEHRSYCS SRARERNFAG EVLGYVTPWN SHGYDVAKVF GSKFTQISPV WLQLKRRGRE MFEITGLHDV DQGWMRAVKK HAKGVRIVPR LLFEDWTYDD FRNVLDSEDE IEELSKTVAQ VAKNQHFDGF VVEVWSQLLS QKHVGLIHML THLAEALHQA RLLVILVIPP AVTPGTDQLG MFTHKEFEQL APILDGFSLM TYDYSTSQQP GPNAPLSWIR ACVQVLDPKS QWRSKILLGL NFYGMDYAAS KDAREPVIGA RYVQTLKDHR PRVVWDSQAA EHFFEYKKNR GGRHVVFYPT LKSLQVRLEL ARELGVGVSI WELGQGLDYF YDLL | |
Greater than 90% as determined by SDS-PAGE. |
68038 | |
20 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |