missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Chitinase domain-containing protein 1
A DNA sequence encoding the Mus musculus (Mouse) Chitinase domain-containing protein 1, was expressed in the hosts and tags indicated.
£476.00 - £1320.00
Specifications
Accession Number | NP_001136153.1 |
---|---|
Gene ID (Entrez) | 68038 |
Name | Chitinase domain-containing protein 1 |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15962808
|
enQuireBio™
QP10054-BA-20UG |
20 μg |
£476.00
20µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15952808
|
enQuireBio™
QP10054-BA-100UG |
100 μg |
£1320.00
100µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15972808
|
enQuireBio™
QP10054-BA-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001136153.1 | |
Chitinase domain-containing protein 1 | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
68038 | |
Research Use Only | |
Chid1 | |
Recombinant Protein | |
Baculovirus | |
TLSKSDAKKA ASKMLLEKTQ FSDKPVQDRG LVVTDIKAED VVLEHRSYCS SRARERNFAG EVLGYVTPWN SHGYDVAKVF GSKFTQISPV WLQLKRRGRE MFEITGLHDV DQGWMRAVKK HAKGVRIVPR LLFEDWTYDD FRNVLDSEDE IEELSKTVAQ VAKNQHFDGF VVEVWSQLLS QKHVGLIHML THLAEALHQA RLLVILVIPP AVTPGTDQLG MFTHKEFEQL APILDGFSLM TYDYSTSQQP GPNAPLSWIR ACVQVLDPKS QWRSKILLGL NFYGMDYAAS KDAREPVIGA RYVQTLKDHR PRVVWDSQAA EHFFEYKKNR GGRHVVFYPT LKSLQVRLEL ARELGVGVSI WELGQGLDYF YDLL | |
Greater than 90% as determined by SDS-PAGE. |