missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse CD74 Protein

A DNA sequence encoding the Mus musculus (Mouse) CD74, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5795-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15967819

  • £560.06 / 100µg
Estimated Shipment: 07-01-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001036070.1
CD74 Protein
100 μg
Not determined.
Extracellular Domain
Mouse
His
Tris-based buffer, 50% glycerol
16149
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Cd74
Recombinant Protein
E. coli
QQQGRLDKLT ITSQNLQLES LRMKLPKSAK PVSQMRMATP LLMRPMSMDN MLLGPVKNVT KYGNMTQDHV MHLLTRSGPL EYPQLKGTFP ENLKHLKNSM DGVNWKIFES WMKQWLLFEM SKNSLEEKKP TEAPPKVLTK CQEEVSHIPA VYPGAFRPKC DENGNYLPLQ CHGSTGYCWC VFPNGTEVPH TKSRGRHNCS EPLDMEDLSS GLGVTRQELG QVTL
Videos
SDS
Documents
Certificates
Special Offers

Special Offers