Learn More
enQuireBio™ Recombinant Mouse CD40L / CD154 / TNFSF5 Protein
Recombinant Protein
Brand:  enQuireBio™ QP5411-100ug
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15946069
  • £476.00 / 100µg
Estimated Shipment: 05-06-2024
to see stock.
Specifications
NP_035746.2
CD40L / CD154 / TNFSF5 Protein
Research Use Only
CD40LG
Recombinant Protein
E. coli
MQRGDEDPQI AAHVVSEANS NAASVLQWAK KGYYTMKSNL VMLENGKQLT VKREGLYYVY TQVTFCSNRE PSSQRPFIVG LWLKPSSGSE RILLKAANTH SSSQLCEQQS VHLGGVFELQ AGASVFVNVT EASQVIHRVG FSSFGLLKL
Greater than 95% as determined by SDS-PAGE.
21947
100 μg
Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Recombinant Mouse CD40L / CD154 / TNFSF5 induced IL-8 production in human PBMCs detected via ELISA (typical ED50 is 1-10 ng/ml). Activity is also measured in HEK-Blue™ CD40L validation criteria in this assay is an ED50 of less than 50 ng/ml.
Mouse
Untagged
10 mM sodium phosphate, 50 mM arginine, pH7.5