missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse ALDOC Protein
A cDNA sequence encoding the ALDOC was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11004-1mg
Additional Details : Weight : 0.01000kg
Specifications
11676 | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMPHSYP ALSAEQKKEL SDIALRIVTP GKGILAADES VGSMAKRLSQ IGVENTEENR RLYRQVLFSA DDRVKKCIGG VIFFHETLYQ KDDNGVPFVR TIQDKGILVG IKVDKGVVPL AGTDGETTTQ GLDGLLERCA QYKKDGADFA KWRCVLKISD RTPSALAILE NANVLARYAS ICQQNGIVPI VEPEILPDGD HDLKRCQYVT EKVLAAVYKA LSDHHVYLEG TLLKPNMVTP GHACPIKYSP EEIAMATVTA LRRTVPPAVP GVTFLSGGQS EEEASLNLNA INRCPLPRPW ALTFSYGRAL QASALNAWRG QRDNAGAATE EFIKRAEMNG LAAQGRYEGS GDGGAAAQSL YIANHAY | |
Greater than 95.0% as determined by SDS-PAGE. |
ALDOC Protein | |
Research Use Only | |
Aldoc | |
Mouse | |
His | |
The ALDOC solution (1 mg/ml) containing Phosphate buffered saline (pH 7.4), 20% glycerol and 1mM DTT. |