missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse AKR1B1 Protein
A cDNA sequence encoding the AKR1B1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10512-1mg
Additional Details : Weight : 0.01000kg
Specifications
AKR1B1 Protein | |
Research Use Only | |
Specific activity is > 500 pmol/min/ug, and is defined as the amount of enzyme that catalyze the reduction of 1.0 pmole DL-glyceraldehyde in the presence of NADPH per minute at pH7.0 at 37C. | |
Mouse | |
His | |
AKR1B1 protein solution (1 mg/ml) containing Phosphate buffered saline (pH 7.4), and 10% glycerol. |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMASHLEL NNGTKMPTLG LGTWKSPPGQ VTEAVKVAID LGYRHIDCAQ VYQNEKEVGV ALQEKLKEQV VKRQDLFIVS KLWCTFHDKS MVKGAFQKTL SDLQLDYLDL YLIHWPTGFK PGPDYFPLDA SGNVIPSDTD FVDTWTAMEQ LVDEGLVKTI GVSNFNPLQI ERILNKPGLK YKPAVNQIEC HPYLTQEKLI EYCHSKGIVV TAYSPLGSPD RPWAKPEDPS LLEDPRIKAI AAKYNKTTAQ VLIRFPIQRN LVVIPKSVTP VRIAENLKVF DFEVSSEDMA TLLSYNRNWR VCALMSCAKH KDYPFHAEV | |
Greater than 95% as determined by SDS-PAGE. |