missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse AK1 Protein
A cDNA sequence encoding the AK1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10509-50ug
Additional Details : Weight : 0.01000kg
Specifications
AK1 Protein | |
Research Use Only | |
Specific activity is > 150 units/mg. One unit will convert 2.0 umoles of ADP to ATP + AMP per minute at pH 7.5 at 37C. | |
Mouse | |
His | |
AK1 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 1mM DTT. |
50 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMGCCVSS EPQEEGGRKT GEKLKKAKII FVVGGPGSGK GTQCEKIVQK YGYTHLSTGD LLRAEVSSGS ERGKKLSAIM EKGELVPLDT VLDMLRDAML AKVDSSNGFL IDGYPREVKQ GEEFEQKIGQ PTLLLYVDAG AETMTQRLLK RGETSGRVDDNEETIKKRLE TYYNATEPVI SFYDKRGIVR KVNAEGTVDT VFSEVCTYLD SLK | |
Greater than 90.0% as determined by SDS-PAGE. |