missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Adck2 Protein

A DNA sequence encoding the Mus musculus (Mouse) Adck2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7772-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16050701

  • £1600.00 / 500µg
Estimated Shipment: 31-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_849204.1
Adck2 Protein
Research Use Only
Adck2
Recombinant Protein
Yeast
FETREPVGSG CVAQVYKAFA SISLLEEDRI WRLGELSAPG TRAVVMQREP FMKDRKPSEN LADEAFLEKL LLPKADLGGS EVGVSQAPWH LPKSDHLIPV AVKVLHPGLL SQVSMDLLLM KIGSKALGLL PGVKWLSLPE IVEEFEKLMV QQTDLRYEAQ NLEHFQHNFQ DMASVKFPTP LRPLITRDIL VETYEESVPV SSYQQAGIPT DLKRKIAQLG INMLLKMIFV DNFVHGDLHP GNILVQGADG VSPSLEMQQQ QVNVCDTLVA TIAPALCPLR LVLLDAGIVA KLQASDLRNF RAVFQAVVMG QGHRVAELML HHAQANECRD VERFKAEMAT LVTQARKNIV TLEKLHVSSL LSSVFKLLMT HKVKLESNFA SIVVAIMVLE GLGRSLDPTL DILEAAKPFL
Greater than 90% as determined by SDS-PAGE.
57869
500 μg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers