missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Activin A / INHBA Protein

Recombinant Protein

Brand:  enQuireBio™ QP5433-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15946119

  • £605.00 / 100µg
Estimated Shipment: 05-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_032406.1
Activin A / INHBA Protein
Research Use Only
INHBA
Recombinant Protein
E. coli
MGLECDGKVN ICCKKQFFVS FKDIGWNDWI IAPSGYHANY CEGECPSHIA GTSGSSLSFH STVINHYRMR GHSPFANLKS CCVPTKLRPM SMLYYDDGQN IIKKDIQNMI VEECGCS
Greater than 95% as determined by SDS-PAGE.
16323
100 μg
Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Shows bioactivity at a concentration of 10 ng/mL in the Cytotoxicity of MPC-11 cells assay. The bioactivity acceptance criteria is set at 1 x 10^5 units/mg.
Mouse
Untagged
Sterile filtered (0.22 um) solution containing 0.1% TFA
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers