missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Monkeypox Virus A35R His-tag Protein
Brand: R&D Systems™ 11285-MX-100
This item is not returnable.
View return policy
Description
Bioassay data are not available.Specifications
NP_536572.1 | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. | |
100 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution. | |
Unconjugated | |
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie® Blue Staining. |
Bioactivity | |
M.W. Observed: 13-21 kDa, under reducing conditions., M.W. Theoretical: 14 kDa | |
Human embryonic kidney cell, HEK293-derived monkeypox virus zaire-96-i-16 A35R protein Arg58-Thr181, with a C-terminal 6-His tag | |
<0.10 EU per 1 μg of the protein by the LAL method. | |
Reconstitute at 500 μg/mL in PBS. | |
Monkeypox Virus A35R |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction