missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mahi Mahi GH Protein

A cDNA sequence encoding the GH was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11962-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15958938

  • £99.15 / 10µg
Estimated Shipment: 09-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

GH Protein
Research Use Only
Recombinant Protein
E. coli
The protein was lyophilized from a concentrated (1 mg/ml) solution with 0.0045mM NaHCO3.
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
ahi Mahi
Untagged
Greater than 97.0% as determined by:(a) Analysis by SEC-HPLC. (b) Analysis by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers