missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant M. tuberculosis MPT51 / MPB51 antigen Protein

A DNA sequence encoding the Mycobacterium tuberculosis (strain CDC 1551 / Oshkosh) MPT51/MPB51 antigen, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7383-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16048570

  • £2480.00 / 1mg
Estimated Shipment: 30-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

WP_003420783.1
1 mg
Not determined.
Full Length
M. tuberculosis
His-SUMO
Tris-based buffer, 50% glycerol
M. tuberculosis MPT51 / MPB51 antigen Protein
Research Use Only
mpt51
Recombinant Protein
E. coli
AEPTAKAAPY ENLMVPSPSM GRDIPVAFLA GGPHAVYLLD AFNAGPDVSN WVTAGNAMNT LAGKGISVVA PAGGAYSMYT NWEQDGSKQW DTFLSAELPD WLAANRGLAP GGHAAVGAAQ GGYGAMALAA FHPDRFGFAG SMSGFLYPSN TTTNGAIAAG MQQFGGVDTN GMWGAPQLGR WKWHDPWVHA SLLAQNNTRV WVWSPTNPGA SDPAAMIGQA AEAMGNSRMF YNQYRSVGGH NGHFDFPASG DNGWGSWAPQ LGAMSGDIVG AIR
Greater than 90% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers