missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant M. tuberculosis MPT51 / MPB51 antigen Protein
A DNA sequence encoding the Mycobacterium tuberculosis (strain CDC 1551 / Oshkosh) MPT51/MPB51 antigen, was expressed in the hosts and tags indicated.
£281.57 - £2480.00
Specifications
Accession Number | WP_003420783.1 |
---|---|
Name | M. tuberculosis MPT51 / MPB51 antigen Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | mpt51 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16028570
|
enQuireBio™
QP7383-EC-100UG |
100 μg |
£715.00
100µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16048570
|
enQuireBio™
QP7383-EC-1MG |
1 mg |
£2480.00
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16038570
|
enQuireBio™
QP7383-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16058570
|
enQuireBio™
QP7383-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16068570
|
enQuireBio™
QP7383-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16078570
|
enQuireBio™
QP7383-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
WP_003420783.1 | |
Research Use Only | |
mpt51 | |
Recombinant Protein | |
E. coli | |
AEPTAKAAPY ENLMVPSPSM GRDIPVAFLA GGPHAVYLLD AFNAGPDVSN WVTAGNAMNT LAGKGISVVA PAGGAYSMYT NWEQDGSKQW DTFLSAELPD WLAANRGLAP GGHAAVGAAQ GGYGAMALAA FHPDRFGFAG SMSGFLYPSN TTTNGAIAAG MQQFGGVDTN GMWGAPQLGR WKWHDPWVHA SLLAQNNTRV WVWSPTNPGA SDPAAMIGQA AEAMGNSRMF YNQYRSVGGH NGHFDFPASG DNGWGSWAPQ LGAMSGDIVG AIR | |
Greater than 90% as determined by SDS-PAGE. |
M. tuberculosis MPT51 / MPB51 antigen Protein | |
Not determined. | |
Full Length | |
M. tuberculosis | |
His-SUMO | |
Tris-based buffer, 50% glycerol |