missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Influenza Vietnam HN31242/2007 Protein
A cDNA sequence encoding the Vietnam HN31242/2007 was constructed and used to recombinantly synthesize the protein.
£99.15 - £1985.00
Specifications
Name | Influenza Vietnam HN31242/2007 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | H |
Product Type | Recombinant Protein |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15975169
|
enQuireBio™
QP13935-2UG |
2 μg |
£99.15
2µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15965169
|
enQuireBio™
QP13935-10UG |
10 μg |
£219.00
10µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15955169
|
enQuireBio™
QP13935-100UG |
100 μg |
£1985.00
100µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Influenza Vietnam HN31242/2007 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Baculovirus | |
ADPMDQICIG YHANNSTEQV DTIMEKNVTV THAQDILEKT HNGKLCDLDG VKPLILRDCS VAGWLLGNPM CDEFINVPEW SYIVEKANPA NDLCYPGNFN DYEELKHLLS RINHFEKIQI IPKSSWSDHE ASSGVSSACP YQGVPSFFRN VVWLIKKNNT YPTIKRSYNN TNQEDLLILW GIHHSNDAAE QTKLYQNPTT YISVGTSTLN QRLVPKIATR SKVNGQSGRM DFFWTILKPN DAINFESNGN FIAPEYAYKI VKKGDSAIMK SEVEYGNCNT KCQTPIGAIN SSMPFHNIHP LTIGECPKYV KSNKLVLATG LRNSPLHHHH HH | |
Greater than 90% as determined by SDS-PAGE. |
Research Use Only | |
H | |
Influenza | |
His | |
The H5N1 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0) and 10% glycerol. |