missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Influenza H3N2 Panama Protein
Recombinant Protein
Brand: enQuireBio™ QP12085-10ug
Additional Details : Weight : 0.01000kg
Specifications
Influenza H3N2 Panama Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
The H3N2 A/Panama/2007/99 solution contains STE, 0.09% sodium azide (NaN3) and 0.005% thimerosal. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Influenza | |
Untagged | |
Greater than 90.0% as determined byAnalysis by SDS-PAGE. |