missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Influenza H1N1 New Caledonia Protein
A cDNA sequence encoding the H1N1 New Caledonia was constructed and used to recombinantly synthesize the protein.
£99.15 - £2205.00
Specifications
Name | Influenza H1N1 New Caledonia Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Influenza |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15979288
|
enQuireBio™
QP12076-2UG |
2 μg |
£99.15
2µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15989288
|
enQuireBio™
QP12076-50UG |
50 μg |
£155.00
50µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15959288
|
enQuireBio™
QP12076-100UG |
100 μg |
£1985.00
100µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15969288
|
enQuireBio™
QP12076-1MG |
1 mg |
£2205.00
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Influenza H1N1 New Caledonia Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Influenza | |
Untagged | |
Greater than 90.0% as determined by: Analysis by SDS-PAGE, HA1 and HA2 bands are observed using SDS-PAGE under reducing conditions. |
Research Use Only | |
Recombinant Protein | |
Influenza H1N1 | |
The Recombinant H1N1 A/New Caledonia/20/99 solution 10mM Sodium phosphate, pH 7.2, 150mM NaCl. |