missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Influenza H1N1 California Protein
A cDNA sequence encoding the H1N1 California was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12075-2ug
Additional Details : Weight : 0.01000kg
Specifications
Influenza H1N1 California Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
The Recombinant H1N1 A/California/04/2009 solution 10mM Sodium phosphate pH 7, 150mM Sodium Chloride, and 0.005% Tween 20. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Influenza | |
Untagged | |
Greater than 90.0% under the conditions that would preserve its biological activity and tertiary structure. |