missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Influenza A H3N2 (Bangkok / 1 / 1979) Hemagglutinin Protein
A DNA sequence encoding the Influenza A virus (strain A/Bangkok/1/1979 H3N2) Influenza A H3N2 (strain A/Bangkok/1/1979) Hemagglutinin, was expressed in the hosts and tags indicated.
£321.05 - £2888.42
Specifications
Name | Influenza A H3N2 (Bangkok / 1 / 1979) Hemagglutinin Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | HA |
Protein Length | Partial (see sequence information for more details). |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16008551
|
enQuireBio™
QP9635-YE-100UG |
100 μg |
£800.00
100µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16048551
|
enQuireBio™
QP9635-YE-500UG |
500 μg |
£1930.00
500µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16018551
|
enQuireBio™
QP9635-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16038551
|
enQuireBio™
QP9635-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16058551
|
enQuireBio™
QP9635-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16028551
|
enQuireBio™
QP9635-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
Influenza A H3N2 (Bangkok / 1 / 1979) Hemagglutinin Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Influenza | |
His | |
Tris-based buffer, 50% glycerol |
Research Use Only | |
HA | |
Recombinant Protein | |
Yeast | |
GIFGAIAGFI ENGWEGMXXG WYGFRHQNSE GTGQAADLKS TQAAIDQING KLNRVIEKTN EKFHQIEKEF SEVEGRIQDL EKYVEDTKID LWSYNAELLV ALENQHTIDL TDSEMNKLFE KTRRQLRENA EDMGNGCFKI YHKCDNACIG SIRNGTYDHD VYRDEALNNR FQIKGVELKX GYKDWILWIS FAISCFLLCV VLLGFIMVXC QKGNIRCNIC I | |
Greater than 90% as determined by SDS-PAGE. |