missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human YWHAE Protein

A cDNA sequence encoding the YWHAE was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13985-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15955319

  • £3965.00 / 1mg
Estimated Shipment: 24-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

7531
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MDDREDLVYQ AKLAEQAERY DEMVESMKKV AGMDVELTVE ERNLLSVAYK NVIGARRASW RIISSIEQKE ENKGGEDKLK MIREYRQMVE TELKLICCDI LDVLDKHLIP AANTGESKVF YYKMKGDYHR YLAEFATGND RKEAAENSLV AYKAASDIAM TELPPTHPIR LGLALNFSVF YYEILNSPDR ACRLAKAAFD DAIAELDTLS EESYKDSTLI MQLLRDNLTL WTSDMQGDGE EQNKEALQDV EDENQ
Greater than 90.0% as determined by SDS-PAGE.
YWHAE Protein
Research Use Only
YWHAE
Human
Untagged
The YWHAE solution (1 mg/1ml) contains phosphate buffered Saline (pH 7.4), 1mM DTT and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers