missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human WLS Protein
A cDNA sequence encoding the WLS was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13962-10ug
Additional Details : Weight : 0.01000kg
Specifications
WLS Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMEMSPWF QFMLFILQLD IAFKLNNQIR ENAEVSMDVS LAYRDDAFAE WTEMAHERVP RKLKCTFTSP KTPEHEGRYY ECDVLPFMEI GSVAHKFYLL NIRLPVNEKK KINVGIGEIK DIRLVGIHQN GGFTK | |
Greater than 90% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
WLS protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0),0.4M Urea and 10% glycerol. |