missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human WIBG Protein

A cDNA sequence encoding the WIBG was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13958-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15945239

  • £155.00 / 10µg
Estimated Shipment: 24-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

WIBG Protein
Research Use Only
Recombinant Protein
E. coli
MEAAGSPAAT ETGKYIASTQ RPDGTWRKQR RVKEGYVPQE EVPVYENKYV KFFKSKPELP PGLSPEATAP VTPSRPEGGE PGLSKTAKRN LKRKEKRRQQ QEKGEAEALS RTLDKVSLEE TAQLPSAPQG SRAAPTAASD QPDSAATTEK AKKIKNLKKK LRQVEELQQR IQAGEVSQPS KEQLEKLARR RALEEELEDL ELGLLEHHHH HH
Greater than 85.0% as determined by SDS-PAGE.
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
WIBG Human solution containing 20mM Tris pH 8.0, 0.1M NaCl and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers