missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human WFDC12 Protein

A cDNA sequence encoding the WFDC12 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13955-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15955229

  • £2975.00 / 1mg
Estimated Shipment: 30-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

WFDC12 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSVKEGIEK AGVCPADNVR CFKSDPPQCH TDQDCLGERK CCYLHCGFKC VIPVKELEEG GNKDEDVSRP YPEPGWEAKC PGSSSTRCPQ K
Greater than 85% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The WFDC12 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 1mM DTT and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers