missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human WBP2 Protein
A cDNA sequence encoding the WBP2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13954-2ug
Additional Details : Weight : 0.01000kg
Specifications
WBP2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMALNKNH SEGGGVIVNN TESILMSYDH VELTFNDMKN VPEAFKGTKK GTVYLTPYRV IFLSKGKDAM QSFMMPFYLM KDCEIKQPVF GANYIKGTVK AEAGGGWEGS ASYKLTFTAG GAIEFGQRML QVASQASRGE VPSGAYGYSY MPSGAYVYPP PVANGMYPCP PGYPYPPPPP EFYPGPPMMD GAMGYVQPPP PPYPGPMEPP VSGPDVPSTP AAEAKAAEAA ASAYYNPGNP HNVYMPTSQP PPPPYYPPED KKTQ | |
Greater than 90% as determined by SDS-PAGE. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The WBP2 solution (0.5 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl, 1mM DTT and 20% glycerol. |