missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human WBP2 Protein
A cDNA sequence encoding the WBP2 was constructed and used to recombinantly synthesize the protein.
£99.15 - £5730.00
Specifications
Name | WBP2 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15945229
|
enQuireBio™
QP13954-2UG |
2 μg |
£99.15
2µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15925229
|
enQuireBio™
QP13954-10UG |
10 μg |
£155.00
10µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15935229
|
enQuireBio™
QP13954-1MG |
1 mg |
£5730.00
1mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
WBP2 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The WBP2 solution (0.5 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl, 1mM DTT and 20% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMALNKNH SEGGGVIVNN TESILMSYDH VELTFNDMKN VPEAFKGTKK GTVYLTPYRV IFLSKGKDAM QSFMMPFYLM KDCEIKQPVF GANYIKGTVK AEAGGGWEGS ASYKLTFTAG GAIEFGQRML QVASQASRGE VPSGAYGYSY MPSGAYVYPP PVANGMYPCP PGYPYPPPPP EFYPGPPMMD GAMGYVQPPP PPYPGPMEPP VSGPDVPSTP AAEAKAAEAA ASAYYNPGNP HNVYMPTSQP PPPPYYPPED KKTQ | |
Greater than 90% as determined by SDS-PAGE. |