missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Vacuolar fusion protein MON1 homolog A Protein

A DNA sequence encoding the Homo sapiens (Human) Vacuolar fusion protein MON1 homolog A, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7799-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16050871

  • £335.00 / 100µg

Estimated Shipment: 26-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001135973.1
Vacuolar fusion protein MON1 homolog A Protein
100 μg
Not determined.
Partial (see sequence information for more details).
Human
His-SUMO
Tris-based buffer, 50% glycerol
84315
Greater than 90% as determined by SDS-PAGE.
Research Use Only
MON1A
Recombinant Protein
E. coli
DGYKVVFVRR SPLVLVAVAR TRQSAQELAQ ELLYIYYQIL SLLTGAQLSH IFQQKQNYDL RRLLSGSERI TDNLLQLMAR DPSFLMGAAR CLPLAAAVRD TVSASLQQAR ARSLVFSILL ARNQLVALVR RKDQFLHPID LHLLFNLISS SSSFREGEAW TPVCLPKFNA AGFFHAHISY LEPDTDLCLL LVSTDREDFF AVSDCRRRFQ ERLRKRGAHL ALREALRTPY YSVAQVGIPD LRHFLYKSKS SGLFTSPEIE APYTSEEEQE RLLGLYQYLH SRAHNASRPL KTIYYTGPNE NLLAWVTGAF ELYMCYSPLG TKASAVSAIH KLMRWIRKEE DRLFILTPLT Y
Videos
SDS
Documents
Certificates