missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human V-type proton ATPase subunit G 1 Protein
A DNA sequence encoding the Homo sapiens (Human) V-type proton ATPase subunit G 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8310-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_004879.1 | |
V-type proton ATPase subunit G 1 Protein | |
Research Use Only | |
ATP6G | |
Recombinant Protein | |
E. coli | |
ASQSQGIQQL LQAEKRAAEK VSEARKRKNR RLKQAKEEAQ AEIEQYRLQR EKEFKAKEAA ALGSRGSCST EVEKETQEKM TILQTYFRQN RDEVLDNLLA FVCDIRPEIH ENYRING | |
Greater than 90% as determined by SDS-PAGE. |
9550 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |