missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Urokinase Protein
A cDNA sequence encoding the Urokinase was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13908-100ug
Additional Details : Weight : 0.01000kg
Specifications
Urokinase Protein | |
Research Use Only | |
Recombinant Protein | |
Human | |
The Urokinase was lyophilized from a concentrated (1 mg/ml) solution containing phosphate buffer. |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
Greater than 94.0% as determined by SDS-PAGE. |