missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Urokinase Protein
A cDNA sequence encoding the Urokinase was constructed and used to recombinantly synthesize the protein.
£99.15 - £550.00
Specifications
Name | Urokinase Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15945089
|
enQuireBio™
QP13908-100UG |
100 μg |
£99.15
100µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15955089
|
enQuireBio™
QP13908-1MG |
1 mg |
£136.00
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15965089
|
enQuireBio™
QP13908-5MG |
5 mg |
£550.00
5mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Urokinase Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
Greater than 94.0% as determined by SDS-PAGE. |
Research Use Only | |
Recombinant Protein | |
Human | |
The Urokinase was lyophilized from a concentrated (1 mg/ml) solution containing phosphate buffer. |