missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human UCHL3 Protein
A cDNA sequence encoding the UCHL3 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10906-20ug
Additional Details : Weight : 0.01000kg
Specifications
UCHL3 Protein | |
Research Use Only | |
Specific activity: >3,000 pmole/min/ug. Measured by the hydrolysis of Ubiquitin-AMC at pH 8.0, at 37C. | |
Human | |
His | |
The UCHL3 protein solution (1 mg/1ml) is formulated in 20mM Tris-HCl buffer (pH 8.0)1mM DTT and 10% glycerol. |
20 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MEGQRWLPLE ANPEVTNQFL KQLGLHPNWQ FVDVYGMDPE LLSMVPRPVC AVLLLFPITE KYEVFRTEEE EKIKSQGQDV TSSVYFMKQT ISNACGTIGL IHAIANNKDK MHFESGSTLK KFLEESVSMS PEERARYLEN YDAIRVTHET SAHEGQTEAP SIDEKVDLHF IALVHVDGHL YELDGRKPFP INHGETSDET LLEDAIEVCK KFMERDPDEL RFNAIALSAA | |
Greater than 95% as determined by SDS-PAGE. |