missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human UBTD2 Protein
A cDNA sequence encoding the UBTD2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13877-20ug
Additional Details : Weight : 0.01000kg
Specifications
UBTD2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMGGCVGA QHDSSGSLNE NSEGTGVALG RNQPLKKEKP KWKSDYPMTD GQLRSKRDEF WDTAPAFEGR KEIWDALKAA AHAFESNDHE LAQAIIDGAN ITLPHGALTE CYDELGNRYQ LPVYCLAPPI NMIEEKSDIE TLDIPEPPPN SGYECQLRLR LSTGKDLKLV VRSTDTVFHM KRRLHAAEGV EPGSQRWFFS GRPLTDKMKF EELKIPKDYV VQVIVSQPVQ NPTPVEN | |
Greater than 95% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The UBTD2 solution (0.5 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl, 1mM DTT and 20% glycerol. |