missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Ubiquitin-like-conjugating enzyme ATG3 Protein
A DNA sequence encoding the Homo sapiens (Human) Ubiquitin-like-conjugating enzyme ATG3, was expressed in the hosts and tags indicated.
£145.46 - £1735.00
Specifications
Accession Number | NP_001265641.1 |
---|---|
Gene ID (Entrez) | 64422 |
Name | Ubiquitin-like-conjugating enzyme ATG3 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15947029
|
enQuireBio™
QP5690-EC-100UG |
100 μg |
£384.00
100µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15967029
|
enQuireBio™
QP5690-EC-1MG |
1 mg |
£1735.00
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15957029
|
enQuireBio™
QP5690-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15977029
|
enQuireBio™
QP5690-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15987029
|
enQuireBio™
QP5690-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15997029
|
enQuireBio™
QP5690-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001265641.1 | |
Ubiquitin-like-conjugating enzyme ATG3 Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
64422 | |
Research Use Only | |
ATG3 | |
Recombinant Protein | |
E. coli | |
MQNVINTVKG KALEVAEYLT PVLKESKFKE TGVITPEEFV AAGDHLVHHC PTWQWATGEE LKVKAYLPTG KQFLVTKNVP CYKRCKQMEY SDELEAIIEE DDGDGGWVDT YHNTGITGIT EAVKEITLEN KDNIRLQDCS ALCEEEEDED EGEAADMEEY EESGLLETDE ATLDTRKIVE ACKAKTDAGG EDAILQTRTY DLYITYDKYY QTPRLWLFGY DEQRQPLTVE HMYEDISQDH VKKTVTIENH PHLPPPPMCS VHPCRHAEVM KKIIETVAEG GGELGVHMYL LIFLKFVQAV IPTIEYDYTR HFTM | |
Greater than 90% as determined by SDS-PAGE. |