missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human UBE2Z Protein
A cDNA sequence encoding the UBE2Z was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13869-1mg
Additional Details : Weight : 0.01000kg
Specifications
UBE2Z Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMSIYKEP PPGMFVVPDT VDMTKIHALI TGPFDTPYEG GFFLFVFRCP PDYPIHPPRV KLMTTGNNTV RFNPNFYRNG KVCLSILGTW TGPAWSPAQS ISSVLISIQS LMTENPYHNE PGFEQERHPG DSKNYNECIR HETIRVAVCD MMEGKCPCPE PLRGVMEKSF LEYYDFYEVA CKDRLHLQGQ TMQDPFGEKR GHFDYQSLLM RLGLIRQKVL ERLHNENAEM DSDSSSSGTE TDLHGSLRV | |
Greater than 95% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The UBE2Z solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 20% glycerol and 1mM DTT. |