missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human UBE2D4 Protein
A DNA sequence encoding the Homo sapiens (Human) UBE2D4, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8267-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_057067.1 | |
UBE2D4 Protein | |
Research Use Only | |
UBE2D4 | |
Recombinant Protein | |
E. coli | |
MALKRIQKEL TDLQRDPPAQ CSAGPVGDDL FHWQATIMGP NDSPYQGGVF FLTIHFPTDY PFKPPKVAFT TKIYHPNINS NGSICLDILR SQWSPALTVS KVLLSICSLL CDPNPDDPLV PEIAHTYKAD REKYNRLARE WTQKYAM | |
Greater than 80% as determined by SDS-PAGE. |
51619 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |