missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human UBE2A Protein
A cDNA sequence encoding the UBE2A was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13852-1mg
Additional Details : Weight : 0.01000kg
Specifications
UBE2A Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MSTPARRRLM RDFKRLQEDP PAGVSGAPSE NNIMVWNAVI FGPEGTPFED GTFKLTIEFT EEYPNKPPTV RFVSKMFHPN VYADGSICLD ILQNRWSPTY DVSSILTSIQ SLLDEPNPNS PANSQAAQLY QENKREYEKR VSAIVEQSWR DC | |
Greater than 95% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
UBE2A Human solution containing 20mM Tris HCl pH 8.0, 1mM DTT, 1mM EDTA and 20% glycerol. |