missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human U2 small nuclear ribonucleoprotein B'' Protein
A DNA sequence encoding the Homo sapiens (Human) U2 small nuclear ribonucleoprotein B'', was expressed in the hosts and tags indicated.
£145.46 - £1735.00
Specifications
Accession Number | NP_003083.1 |
---|---|
Gene ID (Entrez) | 6629 |
Name | U2 small nuclear ribonucleoprotein B'' Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16028540
|
enQuireBio™
QP7374-EC-100UG |
100 μg |
£384.00
100µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16048540
|
enQuireBio™
QP7374-EC-1MG |
1 mg |
£1735.00
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16038540
|
enQuireBio™
QP7374-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16058540
|
enQuireBio™
QP7374-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16068540
|
enQuireBio™
QP7374-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16078540
|
enQuireBio™
QP7374-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_003083.1 | |
U2 small nuclear ribonucleoprotein B'' Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
6629 | |
Research Use Only | |
SNRPB2 | |
Recombinant Protein | |
E. coli | |
MDIRPNHTIY INNMNDKIKK EELKRSLYAL FSQFGHVVDI VALKTMKMRG QAFVIFKELG SSTNALRQLQ GFPFYGKPMR IQYAKTDSDI ISKMRGTFAD KEKKKEKKKA KTVEQTATTT NKKPGQGTPN SANTQGNSTP NPQVPDYPPN YILFLNNLPE ETNEMMLSML FNQFPGFKEV RLVPGRHDIA FVEFENDGQA GAARDALQGF KITPSHAMKI TYAKK | |
Greater than 90% as determined by SDS-PAGE. |