missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human TXNRD3NB Protein

A cDNA sequence encoding the TXNRD3NB was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13843-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15964879

  • £151.37 / 10µg
Estimated Shipment: 11-01-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

645840
Greater than 85.0% as determined by SDS-PAGE.
Research Use Only
TXNRD3NB
Human
His
TXNRD3NB protein solution (0.25 mg/ml) containing 20mM Tris-HCl (pH 8.0), 30% glycerol, 0.15M NaCl and 1mM DTT.
TXNRD3NB Protein
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMRDLSER RLGQPELKAE QQMPLEPVRA RLSVGLACCC SHTTAEASSL EHGDKVFGQG FPSPLEEIKR LLKISRALQA RSVPSTQEKA KCLSGEPGQP EGKGQETYPG PGKVEGKAEP AMRKDDVCPG MKCISG
Videos
SDS
Documents
Certificates
Special Offers

Special Offers