missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human TXN1 Protein

A cDNA sequence encoding the TXN1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10902-25ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15945518

  • £155.00 / 25µg
Estimated Shipment: 31-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

TXN1 Protein
Research Use Only
Specific activity is 7-10 A650/min/mg, obtained by measuring the increase of insulin precipitation in absorbance at 650 nm resulting from the reduction of insulin.
Human
His
TXN1 solution containing 1x PBS pH 7.4.
25 μg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MVKQIESKTA FQEALDAAGD KLVVVDFSAT WCGPCKMIKP FFHSLSEKYS NVIFLEVDVD DCQDVASECE VKCMPTFQFF KKGQKVGEFS GANKEKLEAT INELV
Greater than 95.0% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers