missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TXN1 Protein
A cDNA sequence encoding the TXN1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10902-1mg
Additional Details : Weight : 0.01000kg
Specifications
TXN1 Protein | |
Research Use Only | |
Specific activity is 7-10 A650/min/mg, obtained by measuring the increase of insulin precipitation in absorbance at 650 nm resulting from the reduction of insulin. | |
Human | |
His | |
TXN1 solution containing 1x PBS pH 7.4. |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MVKQIESKTA FQEALDAAGD KLVVVDFSAT WCGPCKMIKP FFHSLSEKYS NVIFLEVDVD DCQDVASECE VKCMPTFQFF KKGQKVGEFS GANKEKLEAT INELV | |
Greater than 95.0% as determined by SDS-PAGE. |