missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human TSSK6 Protein

A cDNA sequence encoding the TSSK6 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13833-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15954849

  • £2975.00 / 1mg
Estimated Shipment: 05-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

TSSK6 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMSGDKLL SELGYKLGRT IGEGSYSKVK VATSKKYKGT VAIKVVDRRR APPDFVNKFL PRELSILRGV RHPHIVHVFE FIEVCNGKLY IVMEAAATDL LQAVQRNGRI PGVQARDLFA QIAGAVRYLH DHHLVHRDLK CENVLLSPDE RRVKLTDFGF GRQAHGYPDL STTYCGSAAY ASPEVLLGIP YDPKKYDVWS MGVVLYVMVT GCMPFDDSDI AGLPRRQKRG VLYPEGLELS ERCKALIAEL LQFSPSARPS AGQVARNCWL RAGDSG
Greater than 80.0% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The TSSK6 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 0.4M Urea.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers