missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Trypsin-2 Protein
A cDNA sequence encoding the Trypsin-2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10899-1mg
Additional Details : Weight : 0.01000kg
Specifications
Trypsin-2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MNLLLILTFV AAAVAAPFDD DDKIVGGYIC EENSVPYQVS LNSGYHFCGG SLISEQWVVS AGHCYKSRIQ VRLGEHNIEV LEGNEQFINA AKIIRHPKYN SRTLDNDILL IKLSSPAVIN SRVSAISLPT APPAAGTESL ISGWGNTLSS GADYPDELQC LDAPVLSQAE CEASYPGKIT NNMFCVGFLE GGKDSCQGDS GGPVVSNGEL QGIVSWGYGC AQKNRPGVYT KVYNYVDWIK DTIAANS | |
Greater than 90% as determined by SDS-PAGE. |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Human | |
Untagged | |
The protein was lyophilized without any additives. |