missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Transcription factor E2F6 Protein
A DNA sequence encoding the Homo sapiens (Human) Transcription factor E2F6, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5958-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001265204.1 | |
Transcription factor E2F6 Protein | |
Research Use Only | |
E2F6 | |
Recombinant Protein | |
E. coli | |
MSQQRPARKL PSLLLDPTEE TVRRRCRDPI NVEGLLPSKI RINLEDNVQY VSMRKALKVK RPRFDVSLVY LTRKFMDLVR SAPGGILDLN KVATKLGVRK RRVYDITNVL DGIDLVEKKS KNHIRWIGSD LSNFGAVPQQ KKLQEELSDL SAMEDALDEL IKDCAQQLFE LTDDKENERL AYVTYQDIHS IQAFHEQIVI AVKAPAETRL DVPAPREDSI TVHIRSTNGP IDVYLCEVEQ GQTSNKRSEG VGTSSSESTH PEGPEEEENP QQSEELLEVS N | |
Greater than 80% as determined by SDS-PAGE. |
1876 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |