missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Transcription factor 7-like 2 Protein
A DNA sequence encoding the Homo sapiens (Human) Transcription factor 7-like 2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8221-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001139746.1 | |
Transcription factor 7-like 2 Protein | |
Research Use Only | |
TCF7L2 | |
Recombinant Protein | |
E. coli | |
MPQLNGGGGD DLGANDELIS FKDEGEQEEK SSENSSAERD LADVKSSLVN ESETNQNSSS DSEAERRPPP RSESFRDKSR ESLEEAAKRQ DGGLFKGPPY PGYPFIMIPD LTSPYLPNGS LSPTARTLHF QSGSTHYSAY KTIEHQIAVQ YLQMKWPLLD VQAGSLQSRQ ALKDARSPSP AHIVSNKVPV VQHPHHVHPL TPLITYSNEH FTPGNPPPHL PADVDPKTGI PRPPHPPDIS PYYPLSPGTV GQIPHPLGWL VPQQGQPVYP ITTGGFRHPY PTALTVNASM SRFPPHMVPP HHTLHTTGIP HPAIVTPTVK QESSQSDVGS LHSSKHQDSK KEEEKKKPHI KKPLNAFMLY MKEMRAKVVA ECTLKESAAI NQILGRRWHA LSREEQAKYY ELARKERQLH MQLYPGWSAR DNYGKKKKRK RDKQPGETNE HSECFLNPCL SLPPIT | |
Greater than 90% as determined by SDS-PAGE. |
6934 | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |