missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Transaldolase Protein

A DNA sequence encoding the Homo sapiens (Human) Transaldolase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8473-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16004811

  • £1688.38 / 1mg

Estimated Shipment
05-10-2020

Please to purchase this item. Need a web account? Register with us today!


Specifications

Specifications

NP_006746.1
Transaldolase Protein
1 mg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
6888
Greater than 90% as determined by SDS-PAGE.
Research Use Only
TAL
Recombinant Protein
E. coli
QRMESALDQL KQFTTVVADT GDFHAIDEYK PQDATTNPSL ILAAAQMPAY QELVEEAIAY GRKLGGSQED QIKNAIDKLF VLFGAEILKK IPGRVSTEVD ARLSFDKDAM VARARRLIEL YKEAGISKDR ILIKLSSTWE GIQAGKELEE QHGIHCNMTL LFSFAMS-QA VACAEAGVTL ISPFVGRILD WHVANTDKKS YEPLEDPGVK SVTKIYNYYK KFSYKTIVMG ASFRNTGEIK ALAGCDFLTI SPKLLGELLQ DNAKLVPVLS AKAAQASDLE KIHLDEKSFR WLHNEDQMAV EKLSDGIRKF AADAVKLERM LTERMFNAEN GK
Videos
Documents