missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human TOP1 70kDa Protein

A cDNA sequence encoding the TOP1 70kDa was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13785-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15944699

  • £155.00 / 20µg
Estimated Shipment: 05-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

7150
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
Baculovirus
MSGDHLHNDS QIEADFRLND SHKHKDKHKD REHRHKEHKK EKDREKSKHS NSEHKDSEKK HKEKEKTKHK DGSSEKHKDK HKDRDKEKRK EEKVRASGDA KIKKEKENGF SSPPQIKDEP EDDGYFVPPK EDIKPLKRPR DEDDADYKPK KIKTEDTKKE KKRKLEEEED GKLKKPKNKD KDKKVPEPDN KKKKPKKEEE QKWKWWEEER YPEGIKWKFL EHKGPVFAPP YEPLPENVKF YYDGKVMKLS PKAEEVATFF AKMLDHEYTT KEIFRKNFFK DWRKEMTNEE KNIITNLSKC DFTQMSQYFK AQTEARKQMS KEEKLKIKEE NEKLLKEYGF CIMDNHKERI ANFKIEPPGL FRGRGNHPKM GMLKRRIMPE DIIINCSKDA KVPSPPPGHK WKEVRHDNKV TWLVSWTENI QGSIKYIMLN PSSRIKGEKD WQKYETARRL KKCVDKIRNQ YREDWKSKEM KVRQRAVALY FIDKLALRAG NEKEEGETAD TVGCCSLRVE HINLHPELDG QEYVVEFDFL GKDSIRYYNK VPVEKRVFKN LQLFMENKQP EDDLFDRLNT GILNKHLQDL MEGLTAKVFR TYNASITLQQ QLKELTAPDE NIPAKILSYN RANRAVAILC NHQRAPPKTF EKSMMNLQTK IDAKKEQLAD ARRDLKSAKA DAKVMKDAKT KKVVESKKKA VQRLEEQLMK LEVQATDREE NKQIALGTSK LNYLDPRITV AWCKKWGVPI EKIYNKTQRE KFAWAIDMAD EDYEF
Greater than 80% as determined by SDS-PAGE.
TOP1 70kDa Protein
Research Use Only
TOP1
Human
His
TOP1 70kDa is supplied in 16mM HEPES buffer pH 8.0, 400mM NaCl, and 20% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers