missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TNFSF10 / TRAIL / APO-2L Protein
Recombinant Protein
£80.80 - £1435.00
Specifications
Accession Number | NP_001177871.1 |
---|---|
Gene ID (Entrez) | 8743 |
Name | TNFSF10 / TRAIL / APO-2L Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15915769
|
enQuireBio™
QP5244-10UG |
10 μg |
£80.80
10µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15945769
|
enQuireBio™
QP5244-50UG |
50 μg |
£191.00
50µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15905769
|
enQuireBio™
QP5244-100UG |
100 μg |
£329.00
100µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15935769
|
enQuireBio™
QP5244-500UG |
500 μg |
£845.00
500µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15925769
|
enQuireBio™
QP5244-1MG |
1 mg |
£1435.00
1mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NP_001177871.1 | |
TNFSF10 / TRAIL / APO-2L Protein | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Human TNFSF10 / TRAIL / APO-2L induces RPMI-8226 cell proliferation (typical ED50 is ? 15 ng/ml). | |
Human | |
Untagged | |
10 mM sodium phosphate, 50 mM sodium chloride, pH 7.5 |
8743 | |
Research Use Only | |
TNFSF10 | |
Recombinant Protein | |
E. coli | |
MRERGPQRVA AHITGTRGRS NTLSSPNSKN EKALGRKINS WESSRSGHSF LSNLHLRNGE LVIHEKGFYY IYSQTYFRFQ EEIKENTKND KQMVQYIYKY TSYPDPILLM KSARNSCWSK DAEYGLYSIY QGGIFELKEN DRIFVSVTNE HLIDMDHEAS FFGAFLVG | |
Greater than 90 as determined by SDS-PAGE. |