missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TNFR1 / CD120a / TNFRSF1A Protein
Recombinant Protein
Brand: enQuireBio™ QP5348-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001056.1 | |
TNFR1 / CD120a / TNFRSF1A Protein | |
Research Use Only | |
TNFRSF1A | |
Recombinant Protein | |
E. coli | |
MDSVCPQGKY IHPQNNSICC TKCHKGTYLY NDCPGPGQDT DCRECESGSF TASENHLRHC LSCSKCRKEM GQVEISSCTV DRDTVCGCKN QYRHYWSENL FQCFNCSLCL NGTVHLSCQE KQNTVCTCHA GFFLRENECV SCSNCKKSLE CTKLCLPQIE N | |
Greater than 95% as determined by SDS-PAGE. |
7132 | |
1 mg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Neutralization of human TNF alpha induced L929 cell cytolysis significantly was induced by less than 100 ng/ml of Human TNFR1 / CD120a / TNFRSF1A which corresponds to an activity of at least 1.0 x 10^4 units/mg. | |
Human | |
Untagged | |
10 mM sodium phosphate, pH 7.5 |