missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TNFR1 / CD120a / TNFRSF1A Protein
Recombinant Protein
£1890.00 - £3085.00
Specifications
Accession Number | NP_001056.1 |
---|---|
Gene ID (Entrez) | 7132 |
Name | TNFR1 / CD120a / TNFRSF1A Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15975939
|
enQuireBio™
QP5348-500UG |
500 μg |
£1890.00
500µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15965939
|
enQuireBio™
QP5348-1MG |
1 mg |
£3085.00
1mg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NP_001056.1 | |
TNFR1 / CD120a / TNFRSF1A Protein | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Neutralization of human TNF alpha induced L929 cell cytolysis significantly was induced by less than 100 ng/ml of Human TNFR1 / CD120a / TNFRSF1A which corresponds to an activity of at least 1.0 x 10^4 units/mg. | |
Human | |
Untagged | |
10 mM sodium phosphate, pH 7.5 |
7132 | |
Research Use Only | |
TNFRSF1A | |
Recombinant Protein | |
E. coli | |
MDSVCPQGKY IHPQNNSICC TKCHKGTYLY NDCPGPGQDT DCRECESGSF TASENHLRHC LSCSKCRKEM GQVEISSCTV DRDTVCGCKN QYRHYWSENL FQCFNCSLCL NGTVHLSCQE KQNTVCTCHA GFFLRENECV SCSNCKKSLE CTKLCLPQIE N | |
Greater than 95% as determined by SDS-PAGE. |