missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human TIMP2 / TIMP-2 Protein

A DNA sequence encoding the Homo sapiens (Human) TIMP2 / TIMP-2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8571-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16005351

  • £1735.00 / 1mg
Estimated Shipment: 03-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_003246.1
TIMP2 / TIMP-2 Protein
Research Use Only
TIMP2
Recombinant Protein
E. coli
SPVHPQQAFC NADVVIRAKA VSEKEVDSGN DIYGNPIKRI QYEIKQIKMF KGPEKDIEFI YTAPSSAVCG VSLDVGGKKE YLIAGKAEGD GKMHITLCDF IVPWDTLSTT QKKSLNHRYQ MGCECKITRC PMIPCYISSP DECLWMDWVT EKNINGHQAK FFACIKRSDG SCAWYRGAAP PKQEFLDIED P
Greater than 90% as determined by SDS-PAGE.
7077
1 mg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers